Imprimer
Anti-CD20 Mouse Monoclonal Antibody (violetFluor® 450) [clone: 2H7]
Fournisseur: Tonbo Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75-0209-T025 75-0209-T100 75-0209-T500
75-0209-T025EA 124 EUR
75-0209-T025 75-0209-T100 75-0209-T500
Anti-CD20 Mouse Monoclonal Antibody (violetFluor® 450) [clone: 2H7]
Anticorps
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human