Imprimer
HPLC columns, LiChroCART®, LiChrospher® 100 RP-18e
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.50170.7084 1.50171.7084 1.50171.7085 1.50173.7084 1.50175.7084 1.50175.7085 1.50177.7084 1.50177.7085 1.50178.7084 1.50178.7085 1.50179.7085 1.50195.7084 1.50195.7085 1.50201.7084 1.50201.7085 1.50228.7084 1.50228.7085 1.50229.7084 1.50229.7085 1.50233.7084 1.50233.7085 1.50234.7084 1.50234.7085 1.50236.7085 1.50302.7085 1.51432.7084 1.51442.7084 1.51442.7085 1.51443.7084 1.51443.7085 1.51444.7084 1.51444.7085 1.51448.7084 1.50174.7084 1.50179.7084 1.50173.7085
1.50170.7084EA 0 EUR
1.50170.7084 1.50171.7084 1.50171.7085 1.50173.7084 1.50175.7084 1.50175.7085 1.50177.7084 1.50177.7085 1.50178.7084 1.50178.7085 1.50179.7085 1.50195.7084 1.50195.7085 1.50201.7084 1.50201.7085 1.50228.7084 1.50228.7085 1.50229.7084 1.50229.7085 1.50233.7084 1.50233.7085 1.50234.7084 1.50234.7085 1.50236.7085 1.50302.7085 1.51432.7084 1.51442.7084 1.51442.7085 1.51443.7084 1.51443.7085 1.51444.7084 1.51444.7085 1.51448.7084 1.50174.7084 1.50179.7084 1.50173.7085
HPLC columns, LiChroCART®, LiChrospher® 100 RP-18e
Colonnes pour chromatographie
Stainless steel LiChroCART® cartridges are available with LiChrospher® sorbents. Ideal for separation of compounds under basic eluents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.