Imprimer
HPLC columns, Hibar®, LiChrospher® 60 RP-select B
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.00423.7093 1.50012.7093 1.50182.7093 1.50183.7093
1.00423.7093EA 0 EUR
1.00423.7093 1.50012.7093 1.50182.7093 1.50183.7093
HPLC columns, Hibar®, LiChrospher® 60 RP-select B
Colonnes pour chromatographie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.