Imprimer
HPLC columns, Hibar®, LiChrospher® 100 RP-18e
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.00423.7084 1.00423.7085 1.00424.7084 1.50012.7085 1.50013.7084 1.50182.7084 1.50182.7085 1.50183.7084 1.50183.7085
1.00423.7084EA 0 EUR
1.00423.7084 1.00423.7085 1.00424.7084 1.50012.7085 1.50013.7084 1.50182.7084 1.50182.7085 1.50183.7084 1.50183.7085
HPLC columns, Hibar®, LiChrospher® 100 RP-18e
Colonnes pour chromatographie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.