Imprimer
Colonnes HPLC, LiChroCART®, Purospher® STAR RP-18e
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.50170.7184 1.50171.7184 1.50171.7185 1.50173.7184 1.50174.7184 1.50175.7184 1.50177.7184 1.50195.7184 1.50229.7184 1.50234.7184 1.51431.7184 1.51432.7184 1.50201.7184 1.50178.7185 1.50201.7185 1.51442.7185 1.50229.7185 1.50236.7184 1.51442.7184 1.50233.7184 1.50233.7185 1.50236.7185 1.51443.7185 1.50302.7184 1.50170.7185 1.50174.7185 1.50175.7185 1.50177.7185 1.51444.7185
1.50170.7184EA 804 EUR
1.50170.7184 1.50171.7184 1.50171.7185 1.50173.7184 1.50174.7184 1.50175.7184 1.50177.7184 1.50195.7184 1.50229.7184 1.50234.7184 1.51431.7184 1.51432.7184 1.50201.7184 1.50178.7185 1.50201.7185 1.51442.7185 1.50229.7185 1.50236.7184 1.51442.7184 1.50233.7184 1.50233.7185 1.50236.7185 1.51443.7185 1.50302.7184 1.50170.7185 1.50174.7185 1.50175.7185 1.50177.7185 1.51444.7185
Colonnes HPLC, LiChroCART®, Purospher® STAR RP-18e
Colonnes pour chromatographie
Les cartouches en acier inoxydable LiChroCART® sont disponibles avec différents types de sorbants Purospher®.
These LiChroCART® cartridges require part number MERC1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number MERC1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: MERC1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and MERC1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.