Imprimer
Test sieves, 400×65 mm
Fournisseur: Retsch GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60.167.000025 60.167.000032 60.167.000038 60.167.000045 60.167.000053 60.167.000063 60.167.000075 60.167.000090 60.167.000106 60.167.000125 60.167.000150 60.167.000180 60.167.000212 60.167.000250 60.167.000300 60.167.000355 60.167.000425 60.167.000500 60.167.000600 60.167.000710 60.167.000850 60.167.001000 60.167.001180 60.167.001400 60.167.001700 60.167.002000 60.167.002360 60.167.002800 60.167.003350 60.167.004000 60.167.004750 60.167.005600 60.167.006300 60.167.006700 60.167.008000 60.167.009500 60.167.011200 60.167.012500 60.167.013200 60.167.016000 60.167.019000 60.167.022400 60.167.025000 60.167.026500 60.167.031500 60.167.037500 60.167.045000 60.167.050000 60.167.053000 60.167.063000 60.167.075000 60.167.090000 60.167.100000 60.167.106000 60.167.125000
510-5561EA 1390 EUR
510-5561 510-5562 510-5563 510-5564 510-5565 510-5566 510-5567 510-5568 510-5569 510-5570 510-5571 510-5572 510-5573 510-5574 510-5575 510-5576 510-5577 510-5578 510-5579 510-5580 510-5581 510-5582 510-5583 510-5584 510-5585 510-5586 510-5587 510-5588 510-5589 510-5590 510-5591 510-5592 510-5593 510-5594 510-5595 510-5596 510-5597 510-5598 510-5599 510-5600 510-5601 510-5602 510-5603 510-5604 510-5605 510-5606 510-5607 510-5608 510-5609 510-5610 510-5611 510-5612 510-5613 510-5614 510-5615
Test sieves, 400×65 mm
Sieves Test Sieves
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labelling comply with standards. The sieves are tested five times and are delivered with test report.
Certifications: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.
Informations de commande: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to website.