Imprimer
Gas detection tubes for water vapour
Fournisseur: Dräger
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8101321 8101781 CH23401 8103061
151-0170EA 153 EUR
151-0170 151-0180 151-3152 DRAE8103061
Gas detection tubes for water vapour
Tubes de détection de gaz
To measure the 'spot concentrations' of water vapour for use with an Accuro® pump.
Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in pipelines or gas installations.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.