Imprimer
VWR®, Tamis de test, 200×32 mm
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
510-0608EA 1130 EUR
510-0608 510-0609 510-0610 510-0611 510-0612 510-0613 510-0614 510-0615 510-0616 510-0617 510-0618 510-0619 510-0620 510-0621 510-0622 510-0623 510-0624 510-0625 510-0626 510-0627 510-0628 510-0629 510-0630 510-0631 510-0632 510-0633 510-0634 510-0635 510-0636 510-0637 510-0638 510-0639 510-0640 510-0641 510-0642 510-0643 510-0644 510-0645 510-0646 510-0647 510-0648 510-0649 510-0650 510-0651 510-0652 510-0653 510-0654 510-0655 510-0656 510-0657 510-0658 510-0659 510-0660 510-0661 510-0662 510-0663 510-0664 510-0665 510-0666 510-0667 510-0668 510-0669 510-0670 510-0671 510-0672 510-0673 510-0674 510-0675 510-0676 510-0677 510-0678
VWR®, Tamis de test, 200×32 mm
Tamis Tamis analytiques
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to www.vwr.com for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informations de livraison: Livrés de série avec un certificat de conformité 2.1 conformément à la norme DIN EN 10204.