Imprimer
Models to practice venipuncture, catheter and other things
Fournisseur: 3B Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1000509 1005171 1005678 1005690 1005806 1000315
BINH1000509EA 0 EUR
BINH1000509 BINH1005171 BINH1005678 BINH1005690 BINH1005806 BINH1000315
Models to practice venipuncture, catheter and other things
Modèles Modèles de biologie humaine
Ideal for training. This revolutionary training arm provides complete venous access for IV therapy and phlebotomy, plus sites for intramuscular and intradermal injections.
  • Valves in the veins can be seen and palpated on the skin surface
  • Hundreds of injections may be performed