Imprimer
Models to show the danger of smoking
Fournisseur: 3B Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1005565 1005578
BINH1005565EA 521 EUR
BINH1005565 BINH1005578
Models to show the danger of smoking
Modèles Modèles de biologie humaine
Smokey Sue dramatically demonstrates the quantity of tar collected in the lungs when one single cigarette is smoked. The tar, normally inhaled directly into the lung, is collected in a transparent tube, and thus shows the quantity of tar which reaches the lung with each cigarette very clearly. Delivered with stand, 3 collection tubes, and carrying bag.
  • Shows effectivly the danger