Imprimer
Hollow fibre filter modules, mPES membrane, MidiKros®
Fournisseur: Spectrum
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MidiKros®
SPECD02-E030-10-SEA 994 EUR
SPECD02-E030-10-S SPECD04-E070-05-N SPECD06-E003-05-S SPECD04-E500-05-N SPECD04-E300-05-N SPECD04-E100-05-N SPECD06-E050-05-N SPECD02-E030-10-N SPECD04-E300-05-S SPECD04-E100-05-S SPECD04-E500-05-S SPECD06-E003-05-N SPECD06-E030-10-S SPECD04-E050-10-S SPECD04-E050-10-N SPECD04-E070-05-S SPECD02-E300-10-S SPECD02-E005-05-S SPECD06-E030-10-N SPECD04-E300-10-N SPECD04-E500-10-N SPECD02-E100-10-S SPECD02-E500-10-S SPECD06-E050-05-S SPECD02-E005-05-N SPECD04-E300-10-S SPECD06-E010-05-S SPECD04-E030-05-S SPECD04-E100-10-N SPECD04-E100-10-S SPECD04-E500-10-S SPECD02-E050-05-S SPECD02-E500-10-N SPECD02-E100-10-N SPECD02-E300-10-N SPECD02-E100-05-S SPECD02-E300-05-S SPECD04-E003-05-S SPECD06-E005-05-N SPECD06-E005-05-S SPECD04-E030-05-N SPECD02-E300-05-N SPECD02-E500-05-S SPECD04-E003-05-N SPECD06-E010-05-N SPECD02-E050-05-N SPECD02-E100-05-N SPECD02-E500-05-N SPECD06-E300-05-S SPECD06-E100-05-N SPECD06-E070-05-S SPECD06-E070-05-N SPECD06-E100-05-S SPECD06-E500-05-S SPECD06-E300-05-N SPECD06-E500-05-N SPECD02-E070-05-N SPECD06-E500-10-N SPECD02-E070-05-S SPECD02-E010-05-S SPECD02-E050-10-N SPECD04-E030-10-N SPECD04-E030-10-S SPECD02-E010-05-N SPECD02-E050-10-S SPECD02-E030-05-N SPECD06-E050-10-N SPECD04-E005-05-S SPECD06-E100-10-S SPECD06-E050-10-S SPECD02-E030-05-S SPECD06-E100-10-N SPECD04-E010-05-S SPECD06-E300-10-S SPECD04-E005-05-N SPECD06-E300-10-N SPECD04-E050-05-S SPECD06-E030-05-S SPECD04-E010-05-N SPECD06-E030-05-N SPECD04-E050-05-N SPECD06-E500-10-S SPECD02-E003-05-N SPECD02-E003-05-S
Hollow fibre filter modules, mPES membrane, MidiKros®
Filtres Systèmes de filtration tangentielle
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • Filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • These filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultrafiltration applications. Ultra-filtration membranes have a high-density skin layer for less fouling and a fairly open outer support structure for increased filtration rates. Micro filtration membranes have a retention defining microporous outer skin and a more open inner layer for increased filtration with higher recoveries. Single-use eliminates the need for costly stainless-steel systems and increases manufacturing flexibility.
Modified polyethersulfone (mPES) is an advanced hydrophilic membrane filtration chemistry that provides higher flux rates for faster processing times, excellent selectivity for separation applications.