Imprimer
Shelving systems, wire, Super Erecta® Pro™
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PR1872NK3 PR2442NK3 PR2454NK3 PR2460NK3 PR2472NK3 PR2424NK3 PR2436NK3 PR2448NK3 PR1860NK3 PR1842NK3 PR1824NK3
METOPR1872NK3EA 0 EUR
METOPR1872NK3 METOPR2442NK3 METOPR2454NK3 METOPR2460NK3 METOPR2472NK3 METOPR2424NK3 METOPR2436NK3 METOPR2448NK3 METOPR1860NK3 METOPR1842NK3 METOPR1824NK3
Shelving systems, wire, Super Erecta® Pro™
Mobilier Étagères
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration. Metroseal 3 with microban provides corrosion resistant finish for wet or high humidity environments with a 12 year warranty against rust and corrosion. Microban antimicrobial is built into the finish to keep the product 'cleaner between cleanings'.
  • Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
  • Versatile construction: by using various accessories, hundreds of shelving configurations become possible
  • Accessibility: shelves can be loaded/unloaded easily from all sides ; this open construction allows use of maximum storage space of cube
  • Adjustability: shelves can be adjusted 25 mm intervals along the entire length of the post
  • Adjustable feet: bolt levelers compensate for surface irregularities
Informations de livraison: Plastic sleeves and wedges are included with each shelf.