Imprimer
Single film frame shippers
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SFFS20101A1202 SFFS20011A5115 SFFS300101B120 SFFS300101B511
ENTESFFS20101A1202EA 0 EUR
ENTESFFS20101A1202 ENTESFFS20011A5115 ENTESFFS300101B120 ENTESFFS300101B511
Single film frame shippers
Matériel de manipulation de wafers Transport de semi-conducteurs
Film frame shippers provide a convenient, safe and ergonomic means to ship and store wafers mounted on film frames. Film frame shippers are designed to protect the frame and to prevent any contact with the wafer surface.
  • Trays stack and snap together conveniently
  • Multiple trays can be shipped or stored together for greater economy
  • Lightweight (1 kg) and easy to carry
  • Parabolic design prevents wafer contact
  • Finger slots for safe wafer extraction
  • Available in ESD protective materials
  • RFID capable to enable monitoring/tracking of product