Imprimer
Colonnes HPLC/UHPLC, kits de développement de méthodes étendues Avantor® ACE®, 1,7 µm
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MDKE-17-1003UEA 1600 EUR
MDKE-17-1003U MDKE-17-1002U MDKE-17-0503U MDKE-17-0502U MDKE-17-0302U MDKE-17-D1002
Colonnes HPLC/UHPLC, kits de développement de méthodes étendues Avantor® ACE®, 1,7 µm
Colonnes pour chromatographie
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Pression max. : 1000 bar
Max. pressure: 1000 bar
Informations de livraison: les kits de développement de méthodes étendues Avantor® ACE® contiennent les colonnes en acier inoxydable Avantor® ACE® SuperC18™, Avantor® ACE® C18-Amide et Avantor® ACE® CN-ES.