Imprimer
Overalls with Bib and Braces, Fristads® PR54-22, Design B, Black (Tall sizes)
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
842940131146 842940131148 842940131150 842940131152 842940131154 842940131156 842940210146 842940210148 842940210150 842940210152 842940210154 842940210156 842940231146 842940231148 842940231150 842940231152 842940231154 842940231156 842940331146 842940331148 842940331150 842940331152 842940331154 842940331156 842940440146 842940440148 842940440150 842940440152 842940440154 842940440156 842940520146 842940520148 842940520150 842940520152 842940520154 842940520156 842940530146 842940530148 842940530150 842940530152 842940530154 842940530156 842940539146 842940539148 842940539150 842940539152 842940539154 842940539156 842940640146 842940640148 842940640150 842940640152 842940640154 842940640156 842940730146 842940730148 842940730150 842940730152 842940730154 842940730156 842940940146 842940940148 842940940150 842940940152 842940940154 842940940156 842940941146 842940941148 842940941150 842940941152 842940941154 842940941156
113-4949EA 0 EUR
113-4949 113-4950 113-4951 113-4952 113-4953 113-4954 113-4937 113-4938 113-4939 113-4940 113-4941 113-4942 113-4955 113-4956 113-4957 113-4958 113-4959 113-4960 113-4961 113-4962 113-4963 113-4964 113-4965 113-4966 113-4967 113-4968 113-4969 113-4970 113-4971 113-4972 113-4907 113-4908 113-4909 113-4910 113-4911 113-4912 113-4919 113-4920 113-4921 113-4922 113-4923 113-4924 113-4913 113-4914 113-4915 113-4916 113-4917 113-4918 113-4925 113-4926 113-4927 113-4928 113-4929 113-4930 113-4931 113-4932 113-4933 113-4934 113-4935 113-4936 113-4973 113-4974 113-4975 113-4976 113-4977 113-4978 113-4943 113-4944 113-4945 113-4946 113-4947 113-4948
Overalls with Bib and Braces, Fristads® PR54-22, Design B, Black (Tall sizes)
Tenue de travail
Overalls made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two chest pockets, one with flap
  • Two front pockets, two back pockets
  • Leg pocket with flap, mobile phone pocket, pen pocket, folding-ruler pocket
  • Adjustable waist, side openings
Choice of many different accent colours.
Weight: 260 g/m²
Basic colour: Black