Imprimer
Horizontal wafer shippers, 200 mm SmartStack®
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HWS200101A61C2 HWS200100-CCPF
ENTEHWS200101A61C2EA 0 EUR
ENTEHWS200101A61C2 ENTEHWS200100-CCPF
Horizontal wafer shippers, 200 mm SmartStack®
Matériel de manipulation de wafers Transport de semi-conducteurs
Horizontal wafer shipper.
  • Packaged, double bagged, heat sealed
  • Automation compatible
Inner cushion
Amine free cushions
Packaged, double bagged, heat sealed
Horizontal wafer shipper kit
Kit consists of: 200mm HWS (1)
Pink inner cushions (13)
Non-woven polyethylene wafer inserts (26)
Corrugated Shipping Box (1)
Outer packing cushions (2)
ESD Bag (1)