Imprimer
Multiple wafer shipping box for 125 mm (5") wafer handling, Ultrapak® series
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H9125-0028 H9125-0029
ENTEH9125-0028EA 0 EUR
ENTEH9125-0028 ENTEH9125-0029
Multiple wafer shipping box for 125 mm (5") wafer handling, Ultrapak® series
Wafer Handling Supplies Wafer Shippers
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.
  • Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
  • Ultrapure polypropylene materials provide a clean shipping environment and assure low levels of condensable inorganics
  • Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
  • Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
  • Assembly overall size: 161×177×160 mm
  • Cassette overall size: 144×153×146 mm