Imprimer
Multiple wafer shipping boxes for 100 mm (4") wafer handling, Ultrapak® series
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H9100-0041 H9100-0042
ENTEH9100-0041EA 0 EUR
ENTEH9100-0041 ENTEH9100-0042
Multiple wafer shipping boxes for 100 mm (4") wafer handling, Ultrapak® series
Matériel de manipulation de wafers Transport de semi-conducteurs
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.
  • Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
  • Ultrapure polypropylene materials provide a clean shipping environment and assure low levels of condensable inorganics
  • Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
En savoir plus