Imprimer
Inserts for trays H 20 series
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H20-001D H20-001B
ENTEH20-001DEA 0 EUR
ENTEH20-001D ENTEH20-001B
Inserts for trays H 20 series
Matériel de manipulation de wafers Plateaux pour semi-conducteurs
Nest between H 20 trays and/or covers.
  • Precut to tray size
  • Inserts provide a softer surface providing protection for more sensitive components
  • Inserts are not intended to limit part migration
  • Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable