Imprimer
Protein digestion kits, trypsin kits, SMART Digest™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60109-101 60109-102 60109-103 60109-101-B 60109-101-MB 60109-102-B 60109-102-MB 60109-103-B 60109-103-MB 60113-101-10PK
555-0087EA 1020 EUR
555-0087 555-0088 555-0089 555-0124 555-0674 HYPE60109-102-B 555-0676 HYPE60109-103-B 555-0675 HYPE60113-101-10PK
Protein digestion kits, trypsin kits, SMART Digest™
Protéines purification Protein Purification Kits
Thermo Scientific™ SMART Digest™ Trypsin Kits are designed to provide a simple to use, fast, highly reproducible and robust process for digestion of proteins in a high throughput format, meeting the needs of biopharmaceutical, bioanalytical and QA/QC environments.
  • Simple process with significantly fewer steps compared to in-solution digests
  • Reduced sample preparation time, typically taking <60 minutes (dependant on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours
  • Higher reproducibility and greater sensitivity than other methods
  • Easily automated for high throughput processing of samples
Informations de livraison: Available as a stand alone kit, or with optional filter plate or SOLAµ™ SPE plate. Bulk resin (-B) and magnetic bulk resin (-MB) are also available separately.