Imprimer
Microplate Sealing Mats
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0030127.960 0030127.978
EPPE0030127.960EA 176 EUR
EPPE0030127.960 EPPE0030127.978
Microplate Sealing Mats
Microplaques Microplaques deepwell
Secure your samples with confidence using Eppendorf® Sealing mats, the ideal solution for incubation and short-term storage needs. These reusable mats can be autoclaved and come in a 'PCR Clean' purity grade (free of RNase, DNase, Human DNA and PCR inhibitors).
  • Sealing mats are reusables and can be autoclaved
  • Suitable for short-term storage