Imprimer
Security units for shelving systems, standard-duty, Super Erecta™
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SEC55S-SD SEC56S-SD
METOSEC55S-SDEA 7240 EUR
METOSEC55S-SD METOSEC56S-SD
Security units for shelving systems, standard-duty, Super Erecta™
Mobilier Étagères
The standard-duty stem caster units protect valuable materials and sensitive items from loss or pilferage.
  • Given height is for unit without casters
  • Ideal for use in wet environments
The heavy-gauge open wire construction keeps contents visible at all time making it easy to check inventory. Shelves can be positioned in 25 mm increments along the entire height of post. Double doors open 270 degrees and can be secured along the sides of the unit.
Informations de commande: The casters and intermediate shelves are not included with base models and must be ordered separately.