Imprimer
Carts
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SXRSBED SXRD43CM3 SXRD76CM3
METOSXRSBEDEA 7960 EUR
METOSXRSBED METOSXRD43CM3 METOSXRD76CM3
Carts
Chariots
The Starsys carts are available in a wide variety of heights and system widths and are ideal for the storage needs in the laboratories. The carts features smooth polymer top/worksurface and are provided with 127 mm polymer swivel casters, the front two casters comes with total-lock toe brake to lock both the wheel and the horn.
  • Easy to clean
  • Interchangeable drawers (76, 152 and 230 mm) with removable totes can be fully extended for easy access
  • Colour-coded drawer pulls are available in an array of choices
  • Label holders available to quickly identify drawer contents