Imprimer
Crosslinkers
Marques : CONSORT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VLBLX-M VLBLX-C VLBLX-L
CONSVLBLX-MEA 4020 EUR
CONSVLBLX-M CONSVLBLX-C CONSVLBLX-L
Crosslinkers
Appareils de réticulation UV
Three models: 254, 312, and 365 nm, each with 5× 8-watt UV tubes.
  • Nine presets for time and energy exposures as well as manual control by the user
  • Timer up to 999,9 minutes
  • Exposure ranges: 0 to 99,99 and 0 to 9,999 J/cm²
  • Storage of the last UV settings in a non-volatile memory
  • Photo-feedback system automatically compensates for output variation and tube aging
  • UV-blocking viewing window
Internal dimensions (W×D×H): 260×330×150 mm
External dimensions (W×D×H): 350×360×310 mm