Imprimer
Cytology funnels, Cytofunnel®, for SHANDON Cytospin® 4
Fournisseur: Epredia
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5991040
SHAN5991040EA 323 EUR
SHAN5991040
Cytology funnels, Cytofunnel®, for SHANDON Cytospin® 4
Entonnoirs Chambres cytologiques
These disposable funnels are used for the shandon Cytofunnel® sample chambers.
  • Pre attached with white filter card
  • Ideal for sample volume up to 0,5 ml