Imprimer
Master mix, BugBuster®
Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71456-3 71456-4
71456-3EA 392 EUR
71456-3 71456-4
Master mix, BugBuster®
Protéines extraction Réactifs pour l'extraction des protéines
BugBuster® master mix combines BugBuster® protein extraction reagent with Benzonase® Nuclease and rLysozyme™ solution in one convenient reagent. It allows for maximum recovery of active soluble protein from both Gram-negative and Gram-positive bacteria. The master mix provides sufficient reagents for protein extraction from 20 g and 100 g cell paste.