Imprimer
Alcometer, recalibration
Fournisseur: SureScreen Diagnostics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SUREAMCALEA 137 EUR
SUREAMCAL
Alcometer, recalibration
Tests pour le dépistage de drogue
After 500 uses the alcometer requires recalibration to ensure that the instrument is reading correctly. It is returned to manufacturer for this service, and turn around is five working days from receipt.