Imprimer
Occupational shoes, lace-up, Uni6, 6781
Fournisseur: Abeba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36781/43 36781/44 36781/35 36781/38 36781/39 36781/47 36781/48 36781/45 36781/46 36781/40 36781/36 36781/37 36781/41 36781/42
114-6107EA 102 EUR
114-6107 114-6108 114-6099 114-6102 114-6103 114-6111 114-6112 114-6109 114-6110 114-6104 114-6100 114-6101 114-6105 114-6106
Occupational shoes, lace-up, Uni6, 6781
Chaussures et bottes
Low shoes made from A-micro, an innovative high tech material, which is especially water resistant and breathable, with non slip sole.
  • Perspiration-absorbent lining with silver fibre
  • Replaceable acc Wave insoles
  • Sanitized® treatment reduces odour and bacterial growth
  • Smooth material is tear resistant, antibacterial and washable
For use in kitchens and food processing areas.
Colour: Black
Certifications: DGUV 112-191; EN 61340; EN 20347:2012; O2, FO, SRC.