Imprimer
Tubing, polyethylene
Fournisseur: Bürkle
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BURK8878-0305EA 57.2 EUR
BURK8878-0305 BURK8878-0406 BURK8878-1014
Tubing, polyethylene
Tuyaux Transfer Tubing
PE, translucent.
  • Temperature resistant from -40 to +95 °C
  • Hardness Shore A 80 ±3
  • Suitable for food
Recognised as safe physiologically, resistant against (non-oxidizing) acids, caustic solutions, salts and a number of solvents, pressure resistant, flexible, impact resistant, light, resistant to compression, up to + 80 °C deformation resistant.