Imprimer
Information signs
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
250567 250566 808990 250555 811628 251678 250550 250552 250150 223904 811618
BRDY250567EA 3.6 EUR
BRDY250567 BRDY250566 BRDY808990 BRDY250555 BRDY811628 BRDY251678 116-0582 116-0583 116-0532 116-0533 BRDY811618
Information signs
Panneaux
White with dark symbols.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm
  • Different sizes- round or square