Imprimer
Real-Time PCR kit for detection of Cronobacter sakazakii
Marques : Q BIOANALYTIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
QB-RT-05-50 QB-RT-05-250 QB-RT-05-1000
710-0672EA 0 EUR
710-0672 710-0671 710-0669
Real-Time PCR kit for detection of Cronobacter sakazakii
Tests microbiologiques Tests PCR
Cronobacter sakazakii (formerly Enterobacter sakazakii) is found naturally in the environment and can survive in dry conditions. It occurs in dry foods, such as powdered infant formula, milk powder and starches as well as in teas. Especially young infants are at high risk developing severe meningitis, if infected. All common Real-Time PCR block cyclers are usable for the test. Each reactions contains an internal amplification control. Therefore, false negative results due to inhibition of the reaction can be excluded. The test included the UNG enzyme to prevent re-amplification of contaminating amplicons. The test can also be used for the confirmation of bacterial colonies. The Real-Time PCR Test allows the detection of the species with high accuracy and is more precise than, cultural methods often are.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certifications: DIN EN ISO 20838 conform detection in food and feed samples from pre-enrichment cultures or for confirmation of bacterial colonies.