Imprimer
Media for primary cells, mammary epithelial cell growth medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-21010 C-21215 C-21210 C-21110
C-21010EA 194 EUR
C-21010 C-21215 C-21210 C-21110
Media for primary cells, mammary epithelial cell growth medium
Milieux de culture cellulaire
PromoCell Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland.
The medium is optimised for primary human cells but can also be used for rat and murine mammary epithelial cells as well as mammalian carcinoma cell lines.
Informations de livraison: PromoCell Mammary Epithelial Cell Growth Medium is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.