Imprimer
Small airway epithelial cell growth medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-21170 C-21070 C-21270 C-21275
C-21170EA 323 EUR
C-21170 C-21070 C-21270 C-21275
Small airway epithelial cell growth medium
Milieux de culture cellulaire
Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth.
The medium is optimised for primary human cells but can also be used for porcine small airway epithelial cells.
Informations de livraison: PromoCell Small Airway Epithelial Cell Growth Medium is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.