Imprimer

Anti-CD14 Mouse Monoclonal Antibody (violetFluor™ 450) [clone: 61D3]

Fournisseur: Tonbo Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

75-0149-T025 75-0149-T100
75-0149-T025EA 73.5 EUR
75-0149-T025 75-0149-T100
Anti-CD14 Mouse Monoclonal Antibody (violetFluor™ 450) [clone: 61D3]
Anticorps
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: violetFluor™ 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR