Imprimer
Nitrile gloves, Solo 999
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34999026 34999027 34999438 34999439
112-5615EA 49.7 EUR
112-5615 112-5616 112-5617 112-5618
Nitrile gloves, Solo 999
Gants
These disposable nitrile gloves provide excellent mechanical protection. They are ideal for use in oily environments.
  • Excellent dexterity thanks to reduced material thickness
  • Chlorinated interior: Easy to put on and to take off
  • Smooth with pebbled fingertips for improved grip
  • Recommended for people sensitised to natural rubber proteins
Typical applications include: Assembly of small oily parts, work with composite materials (resins), laboratory tasks, manufacturing of medicines, pharmaceutical preparation, research, analysis and the handling of precision parts.
Certifications: EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III.