Imprimer
Normal human dermal fibroblasts (NHDF)
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-14031 C-14030
C-14031EA 140 EUR
C-14031 C-14030
Normal human dermal fibroblasts (NHDF)
Cellules
Primary normal human dermal fibroblasts (NHDF) are isolated form the dermis of juvenile foreskin or adult skin from different locations like the face, the breasts, the abdomen, and the thighs.
NHDF can be used for wound healing studies and dermatological research to investigate diseases like scleroderma, fibrosarcoma, fibrosis, xeroderma pigmentosum, and histiocytoma. They are also important for cancer research, tissue regeneration, and tissue engineering studies.
Informations de livraison: Primary normal human dermal fibroblasts can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.