Imprimer
Multiple wafer shippers
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H9150-0037 H9150-0038 H9150-0042
ENTEH9150-0037EA 0 EUR
ENTEH9150-0037 ENTEH9150-0038 ENTEH9150-0042
Multiple wafer shippers
Matériel de manipulation de wafers Transport de semi-conducteurs
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.
  • Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
  • Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
  • Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
En savoir plus