Imprimer
Combined pH electrodes, InPro® 4010
Fournisseur: Mettler - Toledo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
52000512 52000511 52000510
662-0604EA 0 EUR
662-0604 662-0039 METN52000510
Combined pH electrodes, InPro® 4010
Électrodes Électrodes pH
Pressure-resistant, low maintenance electrodes with built-in temperature sensor for universal use in industrial wastewater processes.
  • Low maintenance due to the solid polymer electrode reference system XEROLYT™
  • Open annular junction (no clogging): Electrode is suitable for use in heavily contaminated liquid media
  • Simultaneous pH and temperature measurement for higher performance and enhanced process safety
  • Ruggedly built to withstand rough handling; the shaft is made of strong polymer material
  • Robust, watertight, detachable connector VarioPin (VP) offers excellent shielding against external electrical influences on the measurement signal, and consequently highest process safety
The electrode is particularly suitable for applications in inlet and outflow channels, neutralization tanks, secondary settlers, aeration basins, rainwater catchments, or in pipes and conduits.