Imprimer
Funnels with short stem
Fournisseur: hünersdorff GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
954100 950300 950400 950700 951200
HUEN954100EA 0 EUR
HUEN954100 HUEN950300 HUEN950400 HUEN950700 HUEN951200
Funnels with short stem
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
These food safe HDPE funnels are acid and chemical resistant and are ideal for use in the laboratory, workshop and household purposes.
  • Food-safe
  • Good chemical resistance
  • Good temperature resistance