Imprimer
Media for primary cells, melanocyte culture medium
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-24210 C-24010 C-24110 C-24215
C-24210EA 169 EUR
C-24210 C-24010 C-24110 C-24215
Media for primary cells, melanocyte culture medium
Milieux de culture cellulaire
Melanocyte Culture Medium is serum-free, PMA (Phorbol Myristate Acetate) containing medium optimised for the cultivation of human melanocytes.
The medium is optimised for primary human cells but can also be used for horse, porcine, murine, and rat melanocytes.
Informations de livraison: PromoCell Melanocyte Culture Medium is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.