Imprimer
Bottles, narrow neck, amber glass, Boston type with caps
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
145-4000 145-2360 345-2360 345-4000 245-4000 245-2360
EPSC145-4000EA 176 EUR
EPSC145-4000 EPSC145-2360 EPSC345-2360 EPSC345-4000 EPSC245-4000 EPSC245-2360
Bottles, narrow neck, amber glass, Boston type with caps
Flacons Flacons d'échantillonnage de l'eau
Type III soda-lime amber glass, with screw cap.
  • Excellent chemical resistance
  • Good temperature resistance
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Certifications: Certified to meet EPA performance based specifications for semi-volatile organics, pesticides, PCBs and metals analyses. Available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses, Processed the same way as the certified jugs but without certification documentation, or Unprocessed and ready for your own cleaning procedure.