Imprimer
Sample bottles, narrow neck, round, HDPE, Boston type
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
319-1000 319-0125
EPSC319-1000EA 171 EUR
EPSC319-1000 EPSC319-0125
Sample bottles, narrow neck, round, HDPE, Boston type
Flacons Flacons en plastique
HDPE container with white LDPE-lined polypropylene cap.
  • Narrow mouth for precision pouring
  • Caps assembled onto containers
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each container individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
Recommended for collecting water samples and other low viscosity liquids to be analysed for inorganic trace analytes.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904