Imprimer
Safety spectacles, Jackson Safety® V30 Nemesis
Fournisseur: KIMBERLY CLARK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
25688 25692
KIMB25688EA 9.4 EUR
KIMB25688 KIMB25692
Safety spectacles, Jackson Safety® V30 Nemesis
Lunettes
Frameless ultra-lightweight safety safety glasses with a wrap-around design for added comfort and coverage.
  • 99,9% UVA/UVB protection
  • Sleek, sporty style
  • Soft touch temples for added comfort
  • Every pair includes a neck cordcord
Certifications: EN166 1F; ANSI Z87.1+ 2010