Imprimer
Closures for media bottles
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
342151-0384 342151-0240 342151-0200
NALG342151-0384EA 173 EUR
NALG342151-0384 NALG342151-0240 NALG342151-0200
Closures for media bottles
Capuchons Bouchons pour flacons
Seal PET/PETG square media bottles with Thermo Scientific™ Nalgene™ HDPE closures, which provide a reliable and leakproof† seal. These reliable, high-density polyethylene closures are gamma irradiated to provide a 10-6 Sterility Assurance Level (SAL).
  • Animal Derived Component (ADC) free
Available in 20-415 finish, 24-415 finish and 38-430 finish.
Fit Nalgene PET and PETG media bottles that are sold separately without closures.