Imprimer
Special pH electrodes, IDS (Intelligent Digital Sensors), SenTix®
Fournisseur: WTW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
103743 103751 103752 103753
662-1874EA 411 EUR
662-1874 662-1875 662-1876 662-1877
Special pH electrodes, IDS (Intelligent Digital Sensors), SenTix®
Electrodes pH Electrodes
These special pH electrodes extend the range of the IDS series for enhanced applications in Life Sciences and related applications.
  • Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • Measurement integrity benchmarking of the cells by QSC (Quality Sensor Control)
  • IDS electrodes store their serial number and their calibration records
  • Easy exchange of electrodes thanks to calibration data stored in the sensor
  • GLP support due to automatic documentation of all relevant data
For use with all digital multi-parameter instruments: MultiLine® IDS, inoLab® Multi IDS and pH 3310 IDS.
The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, and more.