Imprimer
ICR swab
Fournisseur: MERCK LIFE SCIENCE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1465290100 1465300001
HEIP1465290100EA 1710 EUR
HEIP1465290100 HEIP1465300001
ICR swab
Écouvillons Écouvillons d'échantillonnage
The ideal swab for microbial monitoring of isolator and clean room surfaces – Requires opening only once for sampling. The ICR Swab is a unique system for environmental monitoring of dry surfaces, such as filling needles, equipment recesses, uneven surfaces and tubing in clean rooms and isolators. It is a qualitative presence/absence test for microbiological contamination. This ‘all-in-one’ heipha ICR Swab System requires opening only once for sampling. All further steps, such as addition of broth, incubation and detection of turbidity, are performed within the closed system, thus minimizing the risk of potential contaminations due to handling.
  • Secure and reliable
  • Convenient
  • Flexible
  • Gamma-irradiated
  • Triple-sleeved
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904