Imprimer
Volumetric flasks, class A, white markings, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.37407 2.37403 2.37404 2.37406 2.37408 2.37409 2.37410 2.37405 2.37402 2.37411 2.37401
BRND2.37407EA 73.1 EUR
BRND2.37407 BRND2.37403 BRND2.37404 BRND2.37406 BRND2.37408 BRND2.37409 BRND2.37410 BRND2.37405 BRND2.37402 BRND2.37411 BRND2.37401
Volumetric flasks, class A, white markings, BLAUBRAND®
Ballons Fioles jaugées
Borosilicate glass 3.3, amber, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • Marquage DE-M
  • Total order from one batch, supplied with batch certificate
*Wide neck.
Certifications: DIN EN ISO 1042