Imprimer
Dropping funnels, cylindrical, with pressure balance
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.895.104 1.895.105 1.895.106
WITG1.895.104EA 264 EUR
WITG1.895.104 WITG1.895.105 WITG1.895.106
Dropping funnels, cylindrical, with pressure balance
Entonnoirs Ampoules de coulée
Borosilicate glass, clear, with PTFE key.
  • Cylindrical, graduated
  • Drip-nozzle with pressure equalizer and screw-thread retaining nut
  • With interchangeable valve made out of PTFE