Imprimer
Replacement impeller for Magna Flex™ flasks
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
356843 356844 356847 356842 356841 356848 356849
734-3145EA 268 EUR
734-3145 734-3146 734-3147 734-3144 734-3143 734-3148 734-3149
Replacement impeller for Magna Flex™ flasks
Agitateurs Axes d'agitation
This unit includes a top cap with liners, glass stirring ball and connecting silicone tubing. This glass impeller assembly is designed for replacement on all Magna Flex™ style flasks.