Imprimer
Volumetric flasks, class A, white markings, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BLAUBRAND®
612-7139EA 59.6 EUR
612-7139 612-7148 612-7149 612-7146 612-7147 612-7144 612-7142 612-7153 612-7143 612-7140 612-7152 612-7141 612-7150
Volumetric flasks, class A, white markings, BLAUBRAND®
Ballons Fioles jaugées
Borosilicate glass 3.3, amber, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • Marquage DE-M
*Wide neck.
Certifications: DIN EN ISO 1042
Informations de commande: Supplied with an individual certificate of calibration.